Order Entry
Netherlands
ContactUsLinkComponent
Anti-SCN4B Rabbit Polyclonal Antibody
Anti-SCN4B Rabbit Polyclonal Antibody
Catalog # ABCAAB219816-50
Supplier:  Abcam
undefined
Anti-SCN4B Rabbit Polyclonal Antibody
Catalog # ABCAAB219816-50
Supplier:  Abcam

Specifications

  • Size:
    50 µl
  • Pack type:
    Vial

Specifications

About this item

Rabbit polyclonal to SCN4B.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: SCN4B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human