Order Entry
Netherlands
ContactUsLinkComponent
 
Anti-CD11c Mouse Monoclonal Antibody [clone: 3.9] (VioletFluor® 450)
  ABCAAB241938-100
 :  Abcam
undefined
Anti-CD11c Mouse Monoclonal Antibody [clone: 3.9] (VioletFluor® 450)
  ABCAAB241938-100
 :  Abcam

 

  • Seq:
    0790
  • Size:
    100 Tests
  • Pack type:
    Vial

 

 

violetFluor™ 450 Mouse monoclonal [3.9] to CD11c.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: CD11c
Clonality: Monoclonal
Clone: 3.9
Conjugation: VioletFluor® 450
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human