Anti-Lysozyme Rabbit Monoclonal Antibody [clone: EPR22270-53]
Supplier: Abcam
Certificates
About this item
Rabbit monoclonal [EPR22270-53] to Lysozyme - BSA and Azide free (Capture).
Type: Primary
Antigen: Lysozyme
Clonality: Monoclonal
Clone: EPR22270-53
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Frequently Bought Together