Order Entry
Netherlands
ContactUsLinkComponent
Rat Neopterin ELISA Kit
Rat Neopterin ELISA Kit
Catalog # ANTIA319480-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  
Rat Neopterin ELISA Kit
Catalog # ANTIA319480-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Competitive
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Rat
  • Target protein:
    Neopterin
  • Description:
    Rat Neopterin ELISA kit
  • Sample type:
    Serum, plasma, tissue homogenates, and other biological fluids
  • Cross reactivity:
    Rat Neopterin ELISA Kit exhibits high specificity and excellent specificity for the detection of rat Neopterin. No significant cross-reactivity or interference between Neopterin and analogues was observed.
  • Detection method:
    Colorimetric
  • Time to Results:
    2 hours
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    31,25 - 2000 pg/ml
  • Storage temperature:
    4 °C
  • Sample volume:
    50 μl
  • Sensitivity:
    18,75 pg/ml
  • Regulatory status:
    RUO
  • Tests per kit:
    96 tests

Specifications

About this item

Rat Neopterin ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of rat Neopterin in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-To-Use ELISA kit
  • Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%

Rat Neopterin ELISA kit employs the competitive enzyme immunoassay technique for the quantitative measurement of rat Neopterin in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with Neopterin antigen. During the incubation, Neopterin present in the samples or standards competes with the fixed amount of immobilised Neopterin for binding sites on the Biotinylated Anti-Neopterin Antibody. The more Neopterin present in a sample or standard, the less Biotinylated Anti-Neopterin Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-Neopterin Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of Neopterin present in each sample or standard. The concentration of Neopterin can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Delivery: This is a Ready-To-Use ELISA kit that contains: Pre-Coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.
Caution: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.