Order Entry
Netherlands
ContactUsLinkComponent
Mouse IL-6 ELISA Kit (High Sensitivity)
Mouse IL-6 ELISA Kit (High Sensitivity)
Catalog # ANTIA323177-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  
Mouse IL-6 ELISA Kit (High Sensitivity)
Catalog # ANTIA323177-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    IL-6
  • Description:
    Mouse IL-6 ELISA kit (high sensitivity)
  • Sample type:
    Serum, plasma, and other biological fluids
  • Cross reactivity:
    No significant cross-reactivity or interference between mouse IL-6 and analogues was observed.
  • Detection method:
    Colorimetric
  • Time to Results:
    3 h 30 min
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    0,78 - 50 pg/ml
  • Storage temperature:
    4 °C
  • Sensitivity:
    0,49 pg/ml
  • Regulatory status:
    RUO
  • Tests per kit:
    96 tests

Specifications

About this item

Mouse IL-6 ELISA kit is a high sensitivity sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse IL-6 in serum, plasma, and other biological fluids.

  • High Sensitivity: Detection at the picogram level!
  • Assay Precision: Intra-Assay: CV <10%, Inter-Assay: CV <10%

Mouse IL-6 ELISA kit employs the sandwich Enzyme-Linked Immunoassay technique for the quantitative measurement of mouse IL-6 in serum, plasma, and other biological fluids. An antibody specific for IL-6 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the IL-6 present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-IL-6 Antibody, which binds the captured IL-6 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Avidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of IL-6 captured in each well. The concentration of IL-6 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Caution: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.