Order Entry
Netherlands
ContactUsLinkComponent
 
Human Synthetic LPHN2 (from HEK293 Cells)
New Product
 
Human Synthetic LPHN2 (from HEK293 Cells)
New Product

 

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Synthetic
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    Lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at –80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
  • Gene ID:
    UniprotID# O95490
  • Reconstitution instructions:
    Reconstitute with distilled sterile water.
  • Carrier-free:
    N
  • Protein synonyms:
    ADGRL2|Adhesion G protein-coupled receptor L2|Calcium-independent alpha-latrotoxin receptor 2|CIRL-2|KIAA0786|Latrophilin homolog 1|Latrophilin-2|LEC1|Lectomedin-1|LPHH1
  • Protein/peptide name:
    LPHN2
  • Molecular weight:
    Full length human LPHN2 protein has a MW of 163,3 kDa.
  • Concentration:
    Reconstitution dependent
  • Formulation:
    Lyophilised from nanodisc solubilisation buffer (20 mM Tris-HCl, 150 mM NaCl, pH 8,0). Normally 5 - 8% trehalose is added as protectants before lyophilisation.
  • Shipping temperature:
    Shipped on blue ice at 4 °C.

 

Frequently Bought Together