Order Entry
Netherlands
ContactUsLinkComponent
Human Synthetic GPRC5A (from HEK293 Cells)
Human Synthetic GPRC5A (from HEK293 Cells)
Supplier:  Antibodies.com
New Product
Certificates
Human Synthetic GPRC5A (from HEK293 Cells)
Supplier:  Antibodies.com
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Synthetic
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
  • Gene ID:
    UniprotID# Q8NFJ5
  • Reconstitution instructions:
    Reconstitute with distilled sterile water
  • Carrier-free:
    N
  • Protein synonyms:
    G-protein coupled receptor family C group 5 member A|GPCR5A|PEIG-1|Phorbol ester induced gene 1|RAI3|RAIG-1|RAIG1|Retinoic acid-induced gene 1 protein|Retinoic acid-induced protein 3
  • Protein/peptide name:
    GPRC5A
  • Molecular weight:
    Full length human GPRC5A protein has a MW of 40,3 kDa
  • Concentration:
    Reconstitution dependent
  • Formulation:
    Lyophilised from nanodisc solubilization buffer (20 mM tris-HCl, 150 mM NaCl, pH 8,0). Normally 5 - 8% trehalose is added as protectants before lyophilisation.
  • Shipping temperature:
    Shipped on blue ice at 4 °C

Specifications

Frequently Bought Together