Order Entry
Netherlands
ContactUsLinkComponent
Human Synthetic NPY1R (from HEK293 Cells)
Human Synthetic NPY1R (from HEK293 Cells)
Supplier:  Antibodies.com
New Product
Certificates
Human Synthetic NPY1R (from HEK293 Cells)
Supplier:  Antibodies.com
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Synthetic
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    Shipped at 4 °C. Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
  • Gene ID:
    UniprotID# P25929
  • Reconstitution instructions:
    Reconstitute with distilled sterile water.
  • Carrier-free:
    N
  • Protein synonyms:
    Neuropeptide Y receptor type 1|NPY1-R|NPYR|NPYY1
  • Protein/peptide name:
    NPY1R
  • Molecular weight:
    Full length human NPY1R protein has a MW of 44,4 kDa.
  • Concentration:
    Reconstitution dependent
  • Formulation:
    Lyophilised from nanodisc solubilisation buffer (20 mM Tris-HCl, 150 mM NaCl, pH 8,0). Normally 5 - 8% trehalose is added as protectants before lyophilisation.
  • Shipping temperature:
    Shipped on blue ice at 4 °C

Specifications

Frequently Bought Together