Recombinant Human PDGFR alpha (from HEK293 cells)
Supplier: Antibodies.com
New Product
Certificates
About this item
Frequently Bought Together
Cryoboxes, Nalgene®
Supplier: Thermo Fisher Scientific
Watermonsternameflessen, steriel
Supplier: VWR Collection
VWR®, Confocale schalen voor microscopie
Supplier: VWR Collection
Cryoboxen, 136×136 mm
Supplier: RATIOLAB
VWR®, Centrifuge Tubes, High Performance, Flat or Plug Cap, PP
Supplier: VWR Collection
VWR® Cryoboxes, Cardboard, 136×136 mm, with Full Labelling
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
Corning® Microplate Deksels
Supplier: Corning
Potje, conisch, met schroefdop, Corning® Gosselin™
Supplier: Corning
Centrifuge tubes, sterile, polypropylene, Corning®
Supplier: Corning
Petrischalen, Star™Dish
Supplier: PHOENIX BIOMEDICAL
VWR®, Cell Culture Flasks, Non Treated or Treated
Supplier: VWR Collection
Eppendorf® Protein LoBind® microcentrifugebuisjes
Supplier: EPPENDORF
VWR® PCR-buisjes, 0,2 ml
Supplier: VWR Collection
Cryoboxen, 133x133 mm
Supplier: RATIOLAB
Monsternamepotjes met snap cap, Corning® Gosselin™
Supplier: Corning
TISSUE CULTURE TREATED 490CM TISSUE CULTURE TREATED ROLLER BOTTLE, VENTED CAP, STERILE 1 * 24 ST
Supplier: MARKET SOURCE PART PROCESS
AriaMx Real-Time PCR systeem
Supplier: AGILENT
Collagen I-coated microplates, Corning® BioCoat®
Supplier: Corning