Human Recombinant IGF1 Receptor (from HEK293 Cells)
Supplier: Antibodies.com
New Product
Certificates
About this item
Partial length Human IGF1 Receptor, expressed in HEK293 cells.
Caution: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:Lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at –80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# P08069
- Reconstitution instructions:Reconstitute with distilled sterile water.
- Carrier-free:N
- Protein synonyms:CD221|IGF-I receptor|Insulin-like growth factor 1 receptor|Insulin-like growth factor I receptor
- Protein/peptide name:IGF1 Receptor
- Purity:>85% by SDS-PAGE and Coomassie blue staining
- Molecular weight:The protein has a predicted molecular mass of 103,7 kDa after removal of the signal peptide. The apparent molecular mass of IGF-1R-His is approximately 130 - 150 kDa due to glycosylation.
- Sequence:IGF-1R (Glu31-Asn932) + 6×His Tag
- Concentration:Reconstitution dependent
- Formulation:Lyophilised from sterile Phosphate Buffered Saline, pH 7,4. Normally 5 - 8% trehalose is added as a protectant before lyophilisation.
- Shipping temperature:Shipped on blue ice at 4 °C.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together