Bestelling invoeren
Netherlands
ContactUsLinkComponent
 
Human Recombinant TIGIT (from HEK293 Cells)
New Product
 
Human Recombinant TIGIT (from HEK293 Cells)
New Product

 

  • Conjugatie:
    Onconjugeerd
  • Protein/peptide type:
    Recombinant
  • Bron:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    Lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at –80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
  • Gen-ID:
    UniprotID# Q495A1
  • Reconstitution instructions:
    Prior to reconstitution, centrifuge the vial at 5000 g for 3 - 5 minutes at room temperature. Reconstitute with appropriate volume of distilled sterile water to bring product to 1 mg/ml concentration. After addition of distilled sterile water, mix by gentle tapping. Note: It is not recommended to vortex or vigorously pipette the sample.
  • Dragervrij:
    N
  • Protein synonyms:
    T-cell immunoreceptor with Ig and ITIM domains|V-set and immunoglobulin domain-containing protein 9|V-set and transmembrane domain-containing protein 3|VSIG9|VSTM3
  • Protein/peptide name:
    TIGIT
  • Zuiverheid:
    >95% by SDS-PAGE and Coomassie blue staining
  • Moleculair gewicht:
    The protein has a predicted molecular mass of 39,13 kDa after removal of the signal peptide. The apparent molecular mass of TIGIT-hFc is approximately 40 - 53 kDa due to glycosylation.
  • Sequentie:
    TIGIT (Met22-Pro141) + hFc (Glu99-Ala330)
  • Concentratie:
    Reconstitution dependent
  • Formulering:
    Lyophilised from sterile phosphate buffered saline, pH 7,4. Normally 5 - 8% Trehalose is added as a protectant before lyophilisation.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Tested applications:
    ELISA, SDS-PAGE

 

Frequently Bought Together