Recombinant Human Interferon alpha 4 (from HEK293 cells)
Catalog # ANTIA317593-50
Supplier: Antibodies.com
New Product
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:Lyophilised: store at −20...−80 °C. Reconstituted: aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# P05013
- Reconstitution instructions:Reconstitute with distilled sterile water.
- Carrier-free:N
- Protein synonyms:IFN-alpha-4|IFN-alpha4a|IFNA4|INFA4|Interferon alpha 4 precursor|Interferon alpha 4B|Interferon alpha 76|Interferon alpha M1|Interferon, alpha 4
- Protein/peptide name:Interferon alpha 4
- Purity:>95% by SDS-PAGE under reducing conditions
- Molecular weight:20,4 KDa
- Concentration:Reconstitution dependent
- Formulation:Lyophilised from a 0,2 µm filtered solution of phosphate buffered Saline, pH 7,4
- Shipping temperature:Shipped on blue ice at 4 °C.
- Tested applications:SDS-PAGE