Order Entry
Netherlands
ContactUsLinkComponent
Human Synthetic NPY5R (from HEK293 Cells)
Human Synthetic NPY5R (from HEK293 Cells)
Supplier:  Antibodies.com
New Product
Certificates
Human Synthetic NPY5R (from HEK293 Cells)
Supplier:  Antibodies.com
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Synthetic
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
  • Gene ID:
    UniprotID# Q15761
  • Reconstitution instructions:
    Reconstitute with distilled sterile water
  • Carrier-free:
    N
  • Protein synonyms:
    Neuropeptide Y receptor type 5|NPY-Y5 receptor|NPY5-R|NPYR5|NPYY5-R
  • Protein/peptide name:
    NPY5R
  • Molecular weight:
    Full length human NPY5R protein has a MW of 50,7 kDa
  • Concentration:
    Reconstitution dependent
  • Formulation:
    Lyophilised from nanodisc solubilization buffer (20 mM tris-HCl, 150 mM NaCl, pH 8,0). Normally 5 - 8% trehalose is added as protectants before lyophilisation.
  • Shipping temperature:
    Shipped on blue ice at 4 °C

Specifications

Frequently Bought Together