Cat Recombinant IL-31RA (from HEK293 Cells)
Supplier: Antibodies.com
New Product
Certificates
About this item
Partial length Feline IL-31RA, expressed in HEK293 cells.
Caution: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Cat
- Storage conditions:Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# UPI001D19966D
- Reconstitution instructions:Reconstitute with distilled sterile water
- Carrier-free:N
- Protein synonyms:CRL3|Cytokine receptor-like 3|GLM-R|Gp130-like monocyte receptor|Gp130-like receptor|GPL|hGLM-R|IL-31 receptor subunit alpha|IL-31R subunit alpha|IL-31R-alpha
- Protein/peptide name:IL-31RA
- Purity:>90% by SDS-PAGE and Coomassie blue staining
- Molecular weight:The protein has a predicted molecular mass of 82,8 kDa after removal of the signal peptide
- Sequence:IL31RA (Val20-Ser516) + mFc (Pro99-Lys330)
- Concentration:Reconstitution dependent
- Formulation:Lyophilised from sterile Phosphate Buffered Saline, pH 7,4. Normally 5 - 8% trehalose is added as protectants before lyophilization.
- Shipping temperature:Shipped on blue ice at 4 °C
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together