Cynomolgus macaque Recombinant ADAM9 (from HEK293 cells)
Supplier: Antibodies.com
New Product
Certificates
About this item
Partial length Cynomolgus macaque ADAM9, expressed in HEK293 cells.
Caution: This product is for research use only. It is not for use in diagnostic or therapeutic procedures.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Cynomolgus macaque
- Storage conditions:Lyophilised: store at −20...−80 °C. Reconstituted: aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# A0A2K5X4X8
- Reconstitution instructions:Reconstitute with distilled sterile water.
- Carrier-free:N
- Protein synonyms:ADAM 9|Cellular disintegrin-related protein|Disintegrin and metalloproteinase domain-containing protein 9|KIAA0021|MCMP|MDC9|Meltrin-gamma|Metalloprotease/disintegrin/cysteine-rich protein 9|MLTNG|Myeloma cell metalloproteinase
- Protein/peptide name:ADAM9
- Purity:>85% by SDS-PAGE and coomassie blue staining
- Molecular weight:The protein has a predicted molecular mass of 749 kDa after removal of the signal peptide. The apparent molecular mass of cADAM9 - His is approximately 55 – 100 kDa due to glycosylation.
- Sequence:ADAM9 (Ala29-Gly698) + 6×His Tag
- Concentration:Reconstitution dependent
- Formulation:Lyophilised from sterile phosphate buffered Saline, pH 7,4, Normally 5 – 8% Trehalose is added as a protectant before lyophilisation
- Shipping temperature:Shipped on blue ice at 4 °C.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together