Recombinant Human DKK1 (from HEK293 cells)
New Product
Frequently Bought Together
Pasteur pipettes, PE
: Alpha Laboratories
VWR®, Straight Sample Containers, with Screw Cap
: VWR Collection
VWR®, Petri Dish, PS
: VWR Collection
Cryoboxes, 133×133 mm
: TENAK
Microplates, 96-well
: Greiner Bio-One
Seals for ALPS™ 50 V heat sealer
: THERMO MATRIX TECHNOLOGIES
Test tubes, with rim
: KARTELL
Cell culture flasks, Nunc™
: Thermo Fisher Scientific
VWR® Blender Bags
: VWR Collection
Media bottles, square, sterile, Nalgene®
: Thermo Fisher Scientific
VWR® Ultra-High Performance Centrifuge Tubes
: VWR Collection
Cryoboxes, 133×133 mm
: RATIOLAB
Serological pipettes, Stripette™
: Corning
VWR®, Cryoboxes, Cardboard, 133×133 mm
: VWR Collection
Glass bottom microplates, SensoPlate™
: Greiner Bio-One