Human Recombinant P2X7 (from HEK293 Cells)
Supplier: Antibodies.com
New Product
Certificates
About this item
Partial length Human P2X7, expressed in HEK293 cells.
Caution: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:Lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at –80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# Q99572
- Reconstitution instructions:Prior to reconstitution, centrifuge the vial at 5000 g for 3 - 5 minutes at room temperature. Reconstitute with appropriate volume of distilled sterile water to bring product to 1 mg/ml concentration. After addition of distilled sterile water, mix by gentle tapping. Note: It is not recommended to vortex or vigorously pipette the sample.
- Carrier-free:N
- Protein synonyms:ATP receptor|P2RX7|P2X purinoceptor 7|P2Z receptor|Purinergic receptor
- Protein/peptide name:P2X7
- Purity:>95% by SDS-PAGE and Coomassie blue staining
- Molecular weight:The protein has a predicted molecular mass of 59,2 kDa after removal of the signal peptide. The apparent molecular mass of hFc-P2RX7 is approximately 55 - 70 due to glycosylation.
- Sequence:hFc (Glu99-Ala330) + P2RX7 (Ser47-Val334)
- Concentration:Reconstitution dependent
- Formulation:Lyophilised from sterile Phosphate Buffered Saline, pH 7,4. Normally 5 - 8% trehalose is added as a protectant before lyophilisation.
- Shipping temperature:Shipped on blue ice at 4 °C.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together