Recombinant Human CD130/gp130 (from HEK293 cells)
New Product
Frequently Bought Together
Media bottles, square, sterile, Nalgene®
: Thermo Fisher Scientific
VWR®, Strips of PCR Tubes and Caps
: VWR Collection
Centrifuge tubes
: Greiner Bio-One
VWR®, Stoppers, PE
: VWR Collection
Lab blender bags, Stomacher®
: Seward (FermionX)
VWR®, Straight Sample Containers, with Screw Cap
: VWR Collection
VWR®, Petri Dish, PS
: VWR Collection
Petri dishes, Corning® Gosselin™
: Corning
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR®, Polystyrene Assay Plates, 96- and 384-Well
: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
: VWR Collection
VIAL 55ML CUSTODY CLEAR 19 1 * 450 items
: Capitol Europe
Stoppers, Pierce-It™
: Simport Scientific
Leit Adhesive Carbon Tabs 12mm (Pack of 100). 1 * 100 items
: MARKET SOURCE PART PROCESS
Storage cryobox, Nalgene®
: Thermo Fisher Scientific
Sample containers, straight, with screw cap, WHEATON®
: DWK Life Sciences