Human Synthetic CCRL2 (from HEK293 cells)
Supplier: Antibodies.com
New Product
Certificates
About this item
Full length Human CCRL2, expressed in HEK293 cells.
Caution: This product is for research use only. It is not for use in diagnostic or therapeutic procedures.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Synthetic
- Source:HEK293 cells
- Species:Human
- Storage conditions:Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# O00421
- Reconstitution instructions:Reconstitute with distilled sterile water
- Carrier-free:N
- Protein synonyms:C-C chemokine receptor-like 2|CCR11|CCR6|Chemokine receptor CCR11|Chemokine receptor X|CKRX|CRAM|HCR|Putative MCP-1 chemokine receptor
- Protein/peptide name:CCRL2
- Molecular weight:Full length human CCRL2 protein has a MW of 39,5 kDa
- Concentration:Reconstitution dependent
- Formulation:Lyophilised from nanodisc solubilisation buffer (20 mM Tris-HCl, 150 mM NaCl, pH 8,0). Normally 5 - 8% trehalose is added as protectants before lyophilisation.
- Shipping temperature:Shipped on blue ice at 4 °C
Specifications
Frequently Bought Together