Recombinant Human CRLR (from HEK293 cells)
Supplier: Antibodies.com
New Product
Certificates
About this item
Frequently Bought Together
Laboratory trays
Supplier: KARTELL
Culture media bottles, square, sterile, Nalgene®
Supplier: Thermo Fisher Scientific
Stoppers, press down, VacuCap®
Supplier: Simport Scientific
CENTRIFUGATION TUBE WITH SCREWCAP 0.5ML 1 * 200 items
Supplier: Roth Carl
Sampling spoon, long handle, disposable Bio
Supplier: BURKLE
Storage tubes, 2D barcoded, Matrix™
Supplier: THERMO MATRIX TECHNOLOGIES
Screw cap microcentrifuge tubes, SuperClear®
Supplier: Labcon
FalconRegistered Mark Round-Bottom Tubes, 14 mL 1 * 500 items
Supplier: STEMCELL Technologies
Transfer pipettes
Supplier: SARSTEDT
Inserts for Brand® cryogenic tubes
Supplier: Brand
Cryoboxes, Cardboard
Supplier: TENAK
Microcentrifuge tubes, 0,5/0,7/1,5/2,0/2,2 ml
Supplier: Greiner Bio-One
VWR®, Blotting Pads, Grade 707
Supplier: VWR Collection
Centrifuge tubes with flat caps, MetalFree®
Supplier: Labcon
VWR®, Coloured Cap Inserts for Cryogenic Vials
Supplier: VWR Collection
BOTTLE PET 250ML SQUARE T/E GREEN CAP 1 * 200 items
Supplier: LAB3 OWN PRODUCTION
Cryoboxes
Supplier: RATIOLAB