Recombinant Human CD276 (from HEK293 cells)
New Product
Frequently Bought Together
VWR®, Strips of PCR Tubes and Caps
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
VWR® Standard Line Transfer Pipettes
: VWR Collection
Cryoboxes, 133×133 mm
: TENAK
VWR® Blender Bags
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
VWR®, Spreaders, L-shaped, Disposable
: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
: VWR Collection
VWR® PCR Tubes, 0,2 ml
: VWR Collection
VWR®, Petri Dish, PS
: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
: VWR Collection