Recombinant Human Ret (from HEK293 cells)
New Product
Frequently Bought Together
Microtubes, with thread
: SARSTEDT
Monsterbuisjes, met schroefdoppen
: BOETTGER PAUL
PCR-platen, 384-well, Armadillo, ABgene®
: Thermo Fisher Scientific
Replicating tool, 96-well, Bel-Blotter™, SP Bel-Art
: Bel-Art Products, a Part of SP
Deepwell Plates 96-well, 500 µl
: EPPENDORF
[EN]ViewPlate-96 Black, Microplate Steri 1 * 50 ST
: PerkinElmer
96- en 384-well platen met optische bodem, Nunc™
: Thermo Fisher Scientific
Monstercontainers met schroefdop
: KARTELL
Filterplaten, 96-wells, AcroPrep™
: Cytiva (Formerly Pall Lab)
Storage Boxes, PP, with Lid
: KOEHLER TECHNISCHE PRODUKTEN
Microplates, 96-well
: Greiner Bio-One
Deep well plates, 96-well, SP Bel-Art
: Bel-Art Products, a Part of SP