Order Entry
Netherlands
ContactUsLinkComponent
 
Human Recombinant IL-15 (from E. coli)
  ANTIA317637-50
New Product
 :  
Human Recombinant IL-15 (from E. coli)
  ANTIA317637-50
New Product
 :  

 

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Seq:
    1202
  • Storage conditions:
    Shipped at 4 °C. Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
  • Gene ID:
    UniprotID# P40933
  • Reconstitution instructions:
    Reconstitute with distilled sterile water.
  • Carrier-free:
    N
  • Protein synonyms:
    IL15|Interleukin-15
  • Protein/peptide name:
    IL-15
  • Purity:
    >95% by SDS-PAGE under reducing conditions
  • Molecular weight:
    12,5 KDa
  • Concentration:
    Reconstitution dependent
  • Formulation:
    Lyophilised from a 0,2 µm filtered solution of 20 mM Phosphate Buffer, 150 mM NaCl, pH 7,0.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Tested applications:
    Cell Proferation Assay, SDS-PAGE

 

 

Partial length Human IL-15, expressed in Escherichia coli, suitable for Cell Proferation Assay, SDS-PAGE.

 : This product is for research use only. It is not for use in diagnostic or therapeutic procedures.