Order Entry
Netherlands
ContactUsLinkComponent
 
Human Synthetic GPR164 (from HEK293 Cells)
New Product
 
Human Synthetic GPR164 (from HEK293 Cells)
New Product

 

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Synthetic
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
  • Gene ID:
    UniprotID# Q8TCB6
  • Reconstitution instructions:
    Reconstitute with distilled sterile water
  • Carrier-free:
    N
  • Protein synonyms:
    D-GPCR|G-protein coupled receptor 164|Olfactory receptor 51E1|Olfactory receptor 52A3|OR51E1|OR51E1P|OR52A3P|POGR|Prostate-overexpressed G protein-coupled receptor
  • Protein/peptide name:
    GPR164
  • Molecular weight:
    Full length human GPR164 protein has a MW of 35,3 kDa
  • Concentration:
    Reconstitution dependent
  • Formulation:
    Lyophilised from nanodisc solubilization buffer (20 mM Tris-HCl, 150 mM NaCl, pH 8,0). Normally 5 - 8% trehalose is added as protectants before lyophilisation.
  • Shipping temperature:
    Shipped on blue ice at 4 °C

 

Frequently Bought Together