Order Entry
Netherlands
ContactUsLinkComponent
Human Synthetic CXCR2 (from HEK293 cells)
Human Synthetic CXCR2 (from HEK293 cells)
Supplier:  Antibodies.com
New Product
Certificates
Human Synthetic CXCR2 (from HEK293 cells)
Supplier:  Antibodies.com
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Synthetic
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
  • Gene ID:
    UniprotID# P25025
  • Reconstitution instructions:
    Reconstitute with distilled sterile water
  • Carrier-free:
    N
  • Protein synonyms:
    C-X-C chemokine receptor type 2|CD182|CDw128b|CXC-R2|CXCR-2|GRO/MGSA receptor|High affinity interleukin-8 receptor B|IL-8 receptor type 2|IL-8R B|IL8RB
  • Protein/peptide name:
    CXCR2
  • Molecular weight:
    Full length human CXCR2 protein has a MW of 40,8 kDa
  • Concentration:
    Reconstitution dependent
  • Formulation:
    Lyophilised from nanodisc solubilisation buffer (20 mM Tris-HCl, 150 mM NaCl, pH 8,0). Normally 5 - 8% Trehalose is added as a protectant before lyophilisation.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Tested applications:
    ELISA, SDS-PAGE

Specifications

Frequently Bought Together