Recombinant Human BAMBI/NMA (from HEK293 cells)
Supplier: Antibodies.com
New Product
Certificates
About this item
Frequently Bought Together
CryoBoxes with hinged lid, for cryovials, 1 to 5 ml, TruCool®
Supplier: BIOCISION
VWR®, Cryo Boxes
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 136x136 mm
Supplier: VWR Collection
Multiwell cell culture plates, Falcon®
Supplier: Corning
Cell culture flasks, Nunc™
Supplier: Thermo Fisher Scientific
Deepwell Plates 96-Well, 2000 µl
Supplier: EPPENDORF
Microcentrifuge tubes, 0,6/1,5/1,7/2,0/5,0 ml, SnapLock, Axygen®
Supplier: Corning
VWR® Cryoboxes, Cardboard, 136×136 mm, with Full Labelling
Supplier: VWR Collection
Strips of 8 PCR tubes with attached caps, 0,2 ml, Amplitube™
Supplier: Simport Scientific
Measuring scoops
Supplier: VITLAB
Cell culture storage bottles, square, PC, Corning®
Supplier: Corning
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
CRYOBOX IN PP 81 POS. H50MM RED 1 * 5 items
Supplier: RATIOLAB
96 MicroWell™ plates, Nunclon™∆
Supplier: Thermo Fisher Scientific
Centrifuge tubes with flat caps, MetalFree®
Supplier: Labcon
NEOBOX 81 PLUS, UNTERTEIL BLAU 1 * 1 ST
Supplier: NEOLAB