Human Recombinant CTLA4 (from HEK293 cells)
Supplier: Antibodies.com
New Product
Certificates
About this item
Partial length Human CTLA4, expressed in HEK293 cells, suitable for ELISA, SDS-PAGE, Flow Cytometry.
Caution: This product is for research use only. It is not for use in diagnostic or therapeutic procedures.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:Lyophilised: store at −20...−80 °C. Reconstituted: aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# P16410
- Reconstitution instructions:Prior to reconstitution, centrifuge the vial at 5000 g for 3 – 5 minutes at room temperature. Reconstitute with appropriate volume of distilled sterile water to bring product to 1 mg/ml concentration. After addition of distilled sterile water, mix by gentle tapping. Note: It is not recommended to vortex or vigorously pipette the sample.
- Carrier-free:N
- Protein synonyms:CD152|CTLA4|Cytotoxic T-lymphocyte protein 4|Cytotoxic T-lymphocyte-associated antigen 4
- Protein/peptide name:CTLA4
- Purity:>95% by SDS-PAGE and coomassie blue staining
- Molecular weight:The protein has a predicted molecular mass of 396 kDa after removal of the signal peptide. The apparent molecular mass of CTLA4 - hFc is approximately 40 – 55 kDa due to glycosylation.
- Sequence:CTLA-4 (Gln36-Ser161) + hFc (Glu99-Ala330)
- Concentration:Reconstitution dependent
- Formulation:Lyophilised from sterile phosphate buffered Saline, pH 7,4, Normally 5 – 8% Trehalose is added as a protectant before lyophilisation
- Shipping temperature:Shipped on blue ice at 4 °C.
- Tested applications:ELISA, SDS-PAGE, Flow Cytometry
Specifications
Frequently Bought Together