Human Synthetic TLR2 (from HEK293 Cells)
Supplier: Antibodies.com
New Product
Certificates
About this item
Full length Human TLR2, expressed in HEK293 cells, suitable for ELISA, SDS-PAGE.
Caution: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Synthetic
- Source:HEK293 cells
- Species:Human
- Storage conditions:Lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at –80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# O60603
- Reconstitution instructions:Reconstitute with distilled sterile water.
- Carrier-free:N
- Protein synonyms:CD282|TIL4|Toll-like receptor 2|Toll/interleukin-1 receptor-like protein 4
- Protein/peptide name:TLR2
- Molecular weight:Full length human TLR2 protein has a MW of 89,8 kDa.
- Concentration:Reconstitution dependent
- Formulation:Lyophilised from nanodisc solubilisation buffer (20 mM Tris-HCl, 150 mM NaCl, pH 8,0). Normally 5 - 8% trehalose is added as a protectant before lyophilisation.
- Shipping temperature:Shipped on blue ice at 4 °C.
- Tested applications:ELISA, SDS-PAGE
Specifications
Frequently Bought Together