Human Recombinant GPCR GPR64 (from HEK293 cells)
New Product
Partial length Human GPCR GPR64, expressed in HEK293 cells.
: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# Q8IZP9
- Reconstitution instructions:Reconstitute with distilled sterile water
- Carrier-free:N
- Protein synonyms:ADGRG2|Adhesion G-protein coupled receptor G2|G-protein coupled receptor 64|He6|Human epididymis-specific protein 6|TM7LN2
- Protein/peptide name:GPCR GPR64
- Purity:>85% by SDS-PAGE and coomassie blue staining
- Molecular weight:The protein has a predicted molecular mass of 64,3 kDa after removal of the signal peptide. The apparent molecular mass of GPR64-His is approximately 100 - 250 kDa due to glycosylation.
- Sequence:GPR64 (Leu38-Ala627) + 6×His Tag
- Concentration:Reconstitution dependent
- Formulation:Lyophilised from sterile Phosphate Buffered Saline, pH 7,4. Normally 5 - 8% Trehalose is added as a protectant before lyophilisation.
- Tested applications:SDS-PAGE
Frequently Bought Together