Human Recombinant VAP1 (from HEK293 Cells)
Supplier: Antibodies.com
New Product
Certificates
About this item
Partial length human VAP1, expressed in HEK293 cells.
Caution: This product is for research use only. It is not for use in diagnostic or therapeutic procedures.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:Lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at –80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# Q16853
- Reconstitution instructions:Reconstitute with distilled sterile water
- Carrier-free:N
- Protein synonyms:Amine oxidase copper-containing 3|Amine oxidase [copper-containing] 3|AOC3|Copper amine oxidase|HPAO|Semicarbazide-sensitive amine oxidase|SSAO|VAP-1|Vascular adhesion protein 1
- Protein/peptide name:VAP1
- Purity:>85% by SDS-PAGE and Coomassie blue staining
- Molecular weight:The protein has a predicted molecular mass of 82,6 kDa after removal of the signal peptide.
- Sequence:6×His tag + AOC3 (Arg28-Asn763)
- Concentration:Reconstitution dependent
- Formulation:Lyophilised from sterile phosphate buffered saline, pH 7,4. Normally 5 - 8% trehalose is added as protectants before lyophilisation.
- Shipping temperature:Shipped on blue ice at 4 °C
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together