
3-(Bromomethyl)-5-chlorobenzo[b]thiophene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1198-51-2
:
Formula:C₉H₆BrClS
MDL Number:MFCD00173769
:261.5700 g/mol
Storage Temperature:Refrigerator
UN:3261
ADR:8,III
:GHS05
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:129.0 to 133.0 deg-C
Specification Test Results
Frequently Bought Together