Methyl-2-oxocyclopentanecarboxylate ≥97.0% (by GC)
: TCI
:10472-24-9
:
:C₇H₁₀O₃
:MFCD00001411
:142.16 g/mol
:111°C
:79°C
:233-962-7
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 97.0 %
Frequently Bought Together