Order Entry
Netherlands
ContactUsLinkComponent
Methyl-2-oxocyclopentanecarboxylate 96%
Methyl-2-oxocyclopentanecarboxylate 96%
Certificates
undefined
Methyl-2-oxocyclopentanecarboxylate 96%

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4550 to 1.4570 (20°C, 589 nm)
  • GC:
    ≥95 %

Specification Test Results

Related Information

Frequently Bought Together