Bis(tri-tert-butylphosphine)palladium(0) ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:53199-31-8
:
Formula:C₂₄H₅₄P₂Pd
MDL Number:MFCD03094580
:511.05 g/mol
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to Yellow to Orange powder to crystal
- Purity (Chelometric Titration):min. 98.0 %
Specification Test Results
Related Information
Frequently Bought Together