Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Fluorene-[D10] 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Fluorene-[D10] 10 µg/ml in cyclohexane, Dr Ehrenstorfer
EHERL20800200CY
:
EHRENSTORFER
:
Fluorene-[D10] 10 µg/ml in cyclohexane, Dr Ehrenstorfer
EHERL20800200CY
:
EHRENSTORFER
:
Pack type:
Glass bottle
Pk:
10 mL
CAS No.:
81103-79-9
Description:
Fluorene-[D10] 10 µg/ml in cyclohexane
Formula:
C₁₃D₁₀
Description:
Fluorene-[D10]