Glycinamide hydrochloride (H-Gly-NH2.HCl) ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1668-10-6
:
Formula:C₂H₇ClN₂O
MDL Number:MFCD00013008
:110.54 g/mol
EINECS:216-789-1
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Argentmetric Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together