Order Entry
Netherlands
ContactUsLinkComponent
Glycinamide hydrochloride (H-Gly-NH2.HCl) 98%
Irritant
Glycinamide hydrochloride (H-Gly-NH2.HCl) 98%
Certificates
undefined
Irritant
Glycinamide hydrochloride (H-Gly-NH2.HCl) 98%

Specification Test Results

  • Appearance:
    White to beige Crystalline powder
  • Infrared spectrum:
    Conforms
  • Titration Argentometric:
    ≥97.5 %
  • Loss on drying:
    ≤1 % (105°C, 4 hrs)
  • TLC:
    ≤1 % Glycine

Specification Test Results

Related Information

Frequently Bought Together