Order Entry
Netherlands
ContactUsLinkComponent
 
Fluorene-2-carbaldehyde ≥95.0% (by titrimetric analysis)
  TCIAF0333-5G
 :  TCI
 :  
undefined
Fluorene-2-carbaldehyde ≥95.0% (by titrimetric analysis)
  TCIAF0333-5G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (Oxime Formation):
    min. 95.0 %
  • Melting point:
    83.0 to 86.0 deg-C
  • Solubility in hot Methanol:
    very faint turbidity

 

 

 :
 :194.23 g/mol
 :250-035-2
 :30084-90-3
 :C₁₄H₁₀O
 :MFCD00001123