Supelco® Organic Reference Standard, cis-9,cis-12-Octadecadienoic acid methyl ester
Catalog # 46950-U
Leverancier: Merck
CAS-nummer:
Specificaties
- Concentration:10 mg/ml in heptane
- Pk:1
- Standards:Certified reference material
- CAS No.:112-63-0
- Description:cis-9,cis-12-Octadecadienoic acid methyl ester
- Storage conditions:–10...–25 °C
- Hazard class description:H225 - H304 - H315 - H336 - H410
- Shipping conditions:STD
- M-Clarity quality level:MQ100
- Flash point:–4 °C
Specificaties
Over dit artikel
- GHS signal: GHS02, GHS08, GHS07, GHS09
Caution: P210 - P233 - P273 - P301 + P310 - P303 + P361 + P353 - P331