Order Entry
Netherlands
ContactUsLinkComponent
 
 
 
2-(Trimethylsilyloxy)-1,3-cyclohexadiene ≥94.0% (by GC)
  TCIAT2630-5G
 :  TCI
 :  
undefined
 
 
2-(Trimethylsilyloxy)-1,3-cyclohexadiene ≥94.0% (by GC)
  TCIAT2630-5G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 94.0 %

 

 

 :
 :60°C
 :47°C
 :Refrigerator
 :1993
 :3,III
 :GHS02
 :GHS07
 :54781-19-0
 :C₉H₁₆OSi
 :MFCD00274202