
4'-Acetylbenzo-15-crown-5 (4'-acetylbenzo-1,4,7,10,13-pentaoxacyclopentadecane) ≥97.0% (by GC)
TCIAA1603-1G
: TCI
:
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:95.0 to 99.0 deg-C
:
:GHS07
:41757-95-3
:C₁₆H₂₂O₆
:MFCD00188564