
4-tert-Butylcyclohexyl acetate (mixture of cis and trans isomers) cis- and trans-mixture ≥96.0% (by GC)
: TCI
:32210-23-4
:
:C₁₂H₂₂O₂
:MFCD00019354
:102°C
:250-954-9
:GHS07
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 96.0% (total of isomer)
Frequently Bought Together