Order Entry
Netherlands
ContactUsLinkComponent
3,4-Dimethylcyclohexanone mixture of isomers ≥98.0% (by GC)
Flammable
3,4-Dimethylcyclohexanone mixture of isomers ≥98.0% (by GC)
Catalog # TCIAD1281-10ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
3,4-Dimethylcyclohexanone mixture of isomers ≥98.0% (by GC)
Catalog # TCIAD1281-10ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • Seq:
    2275

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 98.0 % (total of isomers)

Specification Test Results

About this item

 :
UN:1224
ADR:3,III
 :GHS02
CAS Number:5465-09-8
Formula:C₈H₁₄O
MDL Number:MFCD00045552