
3,5-Dibromo-2-pyridone ≥98.0% (by GC, titration analysis)
Catalog # TCIAD2918-5G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:207.0 to 212.0 deg-C
Specification Test Results
About this item
:
:252.89 g/mol
:GHS07
CAS Number:13472-81-6
Formula:C₅H₃Br₂NO
MDL Number:MFCD00023472