(4-Nitrophenyl)-α-D-glucopyranoside ≥98.0% (by HPLC) substrate for alpha-D-Glucosidase
Supplier: TCI
Certificates
About this item
CAS Number:3767-28-0
:
Formula:C₁₂H₁₅NO₈
MDL Number:MFCD00064088
:301.25 g/mol
Storage Temperature:Refrigerator
EINECS:223-189-3
Specification Test Results
- Purity (HPLC):min. 98.0 area%
- Specific rotation [a]20/D:+215 to +222 deg (C=0.5, H2O)
Specification Test Results
Frequently Bought Together