
3-(2-Benzimidazolyl)-7-(diethylamino)coumarin ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:27425-55-4
:
Formula:C₂₀H₁₉N₃O₂
MDL Number:MFCD00051348
:333.3838 g/mol
EINECS:248-451-4
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:240.0 to 244.0 deg-C
- Molar extinction coefficient:min. 48000 (THF, 429 to 433nm)
Specification Test Results
Frequently Bought Together