
(4-Nitrobenzyl)triphenylphosphonium chloride ≥98.0% (by HPLC, titration analysis)
TCIAN0701-25G
: TCI
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
:
:478.33 g/mol
:275°C
:220-444-0
:GHS07
:2767-70-6
:C₂₅H₂₁BrNO₂P
:MFCD00032108