Order Entry
Netherlands
ContactUsLinkComponent
2'-Deoxyadenosine, anhydrous ≥98.0% (by HPLC, titration analysis)
2'-Deoxyadenosine, anhydrous ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAD4137-5G
Supplier:  TCI
CAS Number:  
undefined
2'-Deoxyadenosine, anhydrous ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAD4137-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    188.0 to 192.0 deg-C
  • Specific rotation [a]20/D:
    -25.0 to -28.0 deg (C=0.5, H2O)

Specification Test Results

About this item

 :
 :251.24 g/mol
Storage Temperature:Refrigerator
EINECS:213-488-7
CAS Number:958-09-8
 :MFCD00005754
Formula:C₁₀H₁₃N₅O₃