
(Ethoxycarbonylmethyl)triphenylphosphonium bromide ≥97.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1530-45-6
:
Formula:C₂₂H₂₂BrO₂P
MDL Number:MFCD00011835
:429.30 g/mol
EINECS:216-230-1
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (Nonaqueous Titration):min. 97.0 %
Specification Test Results
Related Information
Frequently Bought Together