
Cyclopentane-1,1-diacetic acid ≥98.0% (by GC, titration analysis)
Catalog # TCIAC0838-5G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:177.0 to 182.0 deg-C
- Solubility in Methanol:within almost transparency
Specification Test Results
About this item
:
:186.21 g/mol
EINECS:240-761-8
:GHS07
CAS Number:16713-66-9
Formula:C₉H₁₄O₄
MDL Number:MFCD00001388